1,3-Dimethylimidazolium chloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,3-Dimethyl-1H-imidazol-3-ium chloride
A255596-1G
A255596-5G
A255596-25G
A255596-10G
A255596-1G
77304-652EA
7
USD
77304-652
103852-058
103812-196
76976-798
76976-796
1,3-Dimethylimidazolium chloride 97%
1,3-Dimethylimidazolium chloride
Caution: for Research Use Only
Formula:
C₅H₉ClN₂ MW: 132.5900 g/mol Melting Pt: 125 °C Storage Temperature: Ambient |
MDL Number:
MFCD08276366 CAS Number: 79917-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...