1,3-Dichloro-2-iodobenzene 98%

Supplier: Ambeed
Warning

A263931-5G A263931-100G A263931-1G A263931-10G A263931-25G A263931-500G A263931-5G A263931-1G
77092-184EA 20.6 USD
77092-184 77092-186 77098-878 77304-880 77098-882 77098-880 103852-538 103812-646
1,3-Dichloro-2-iodobenzene 98%
1,3-Dichloro-2-iodobenzene

Caution: for Research Use Only

Formula: C₆H₃Cl₂I
MW: 272.90 g/mol
Boiling Pt: 268.3 °C (760 mm Hg)
Melting Pt: 64…67 °C
Flash Pt: 116.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00060658
CAS Number: 19230-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR