1,2-Dimethyl-3-propylimidazolium iodide 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,2-Dimethyl-3-propyl-1H-imidazol-3-ium iodide
A109035-25G
A109035-1G
A109035-5G
A109035-1G
76821-690EA
103
USD
76821-690
103823-222
77310-050
103797-434
1,2-Dimethyl-3-propylimidazolium iodide 95%
1,2-Dimethyl-3-propylimidazolium iodide
Caution: for Research Use Only
Formula:
C₈H₁₅IN₂ MW: 266.1226 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15072160 CAS Number: 218151-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...