1,2-Diiodoethane 97%

Supplier: Ambeed
Warning

A541330-5G A541330-100G A541330-500G A541330-25G A541330-5G A541330-1G A541330-1G
77197-400EA 123 USD
77197-400 76843-804 76843-806 76843-808 103828-634 77197-402 103839-754
1,2-Diiodoethane 97%
1,2-Diiodoethane

Caution: for Research Use Only

Formula: C₂H₄I₂
MW: 281.86 g/mol
Boiling Pt: 206 °C (760 mm Hg)
Melting Pt: 80…83 °C
Flash Pt: 100.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001093
CAS Number: 624-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR