3'-Fluoro-2'-methoxyacetophenone 95%

Supplier: Ambeed

Warning

Synonyms: 1-(3-Fluoro-2-methoxyphenyl)ethanone

A129355-1G A129355-5G
103801-270EA 46 USD
103801-270 103832-154
3'-Fluoro-2'-methoxyacetophenone 95%
3'-Fluoro-2'-methoxyacetophenone

Caution: Sealed in dry, at room temperature.

Formula: C₉H₉FO₂
MW: 168.17 g/mol
Boiling Pt: 185 °C (760 mm Hg)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD12545933
CAS Number: 295779-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR