1-(3,5-Dichloro-4-pyridinyl)piperazine 98%

Warning

A807428-5G
103841-076EA 198.46 USD
103841-076
1-(3,5-Dichloro-4-pyridinyl)piperazine 98%
1-(3,5-Dichloro-4-pyridinyl)piperazine

Caution: Keep in a dark place, inert atmosphere, room temperature

Formula: C₉H₁₁Cl₂N₃
MW: 232.11 g/mol
Boiling Pt: 347.1 °C (760 mm Hg)
Flash Pt: 163.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00221400
CAS Number: 175277-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR