3'-Benzyloxyacetophenone 98%

Supplier: Ambeed

Warning

Synonyms: 1-(3-(Benzyloxy)phenyl)ethanone1-[3-(Benzyloxy)phenyl]ethan-1-one

A347637-1G A347637-5G
103825-976EA 7 USD
103825-976 103836-962
3'-Benzyloxyacetophenone 98%
3'-Benzyloxyacetophenone

Caution: Sealed in dry, at room temperature.

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Boiling Pt: 354 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00026221
CAS Number: 34068-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR