2',4'-Bis(trifluoromethyl)acetophenone 98%

Warning

Synonyms: 1-[2,4-Bis(trifluoromethyl)phenyl]ethan-1-one1-(2,4-Bis(trifluoromethyl)phenyl)ethanone

A191759-1G A191759-5G A191759-1G
103847-002EA 129.23 USD
103847-002 103816-400
2',4'-Bis(trifluoromethyl)acetophenone 98%
2',4'-Bis(trifluoromethyl)acetophenone

Caution: for Research Use Only

Formula: C₁₀H₆F₆O
MW: 256.15 g/mol
Boiling Pt: 211 °C (760 mm Hg)
Flash Pt: 78.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00042495
CAS Number: 237069-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR