(S)-(-)-4-Benzyl-2-oxazolidinone 98%

Supplier: AMBEED, INC
Warning

Synonyms: (S)-4-Benzyloxazolidin-2-one(S)-4-Benzyl-2-oxazolidinone

A104875-5G A104875-10G A104875-1000G A104875-25G A104875-100G A104875-500G
76671-486EA 8 USD
76671-486 76671-484 103807-920 76671-482 76671-490 76671-488
(S)-(-)-4-Benzyl-2-oxazolidinone 98%
(S)-(-)-4-Benzyl-2-oxazolidinone

Caution: for Research Use Only

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Boiling Pt: 398.8 °C (760 mm Hg)
Melting Pt: 86…88 °C
Flash Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00064496
CAS Number: 90719-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR