(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine 97%

Supplier: Ambeed

Warning

Synonyms: (S)-1,2,3,4-Tetrahydronaphthalen-1-amine

A215372-1G A215372-5G
103818-482EA 8 USD
103818-482 103849-186
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine 97%
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine

Caution: Keep in a dark place, inert atmosphere, room temperature

Formula: C₁₀H₁₃N
MW: 147.22 g/mol
Boiling Pt: 249.6 °C (760 mm Hg)
Melting Pt: <…20 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00671630
CAS Number: 23357-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR