(R)-(+)-1-Boc-3-aminopyrrolidine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(R)-tert-Butyl 3-aminopyrrolidine-1-carboxylate
, (3R)-(+)-1-Boc-3-aminopyrrolidine
, (3R)-(+)-3-Amino-1-Boc-pyrrolidine
, (3R)-(+)-3-Amino-1-(tert-butoxycarbonyl)pyrrolidine
, (3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine
A182109-1G
A182109-1G
A182109-5G
A182109-10G
A182109-5G
A182109-25G
103846-052EA
25.38
USD
103846-052
103807-684
77083-588
77301-768
77083-586
77191-716
(R)-(+)-1-Boc-3-aminopyrrolidine 98%
(R)-(+)-1-Boc-3-aminopyrrolidine
Caution: for Research Use Only
Formula:
C₉H₁₈N₂O₂ MW: 186.25 g/mol Boiling Pt: 243…244 °C (760 mm Hg) Flash Pt: 91.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03419272 CAS Number: 147081-49-0 UN: 2922 ADR: 8,6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...