D-(+)-Lactamide ≥95%

Supplier: Ambeed

Warning

Synonyms: (R)-2-Hydroxypropanamide

A277523-5G A277523-1G
103813-356EA 9 USD
103813-356 103853-290
D-(+)-Lactamide ≥95%
D(+)-Lactamide

Caution: Inert atmosphere, 2…8 °C.

Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 281.9 °C (760 mm Hg)
Melting Pt: 50…53 °C
Flash Pt: 124.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00210310
CAS Number: 598-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR