(R)-1-Boc-3-Isopropylpiperazine ≥95%

Supplier: Ambeed

Warning

A102189-5G A102189-1G
103796-332EA 193 USD
103796-332 103822-026
(R)-1-Boc-3-Isopropylpiperazine ≥95%
(R)-1-Boc-3-isopropylpiperazine

Caution: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₁₂H₂₄N₂O₂
MW: 228.3300 g/mol
Boiling Pt: 298.4 °C (760 mm Hg)
Flash Pt: 134.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07772113
CAS Number: 928025-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR