5-Chloro-2-methylphenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(5-Chloro-2-methylphenyl)boronic acid
, 5-Chloro-2-methylbenzeneboronic acid
A111489-5G
A111489-1G
A111489-5G
A111489-1G
77310-468EA
17
USD
77310-468
103797-846
77284-124
103823-666
5-Chloro-2-methylphenylboronic acid 97%
5-Chloro-2-methylphenylboronic acid
Caution: for Research Use Only
Formula:
C₇H₈BClO₂ MW: 170.40 g/mol Boiling Pt: 328 °C (760 mm Hg) Melting Pt: 162…166 °C Flash Pt: 152.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411939 CAS Number: 148839-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...