4-Aminophenylboronic acid hydrochloride 95%

Supplier: Ambeed

Warning

Synonyms: (4-Aminophenyl)boronic acid hydrochloride4-Aminobenzeneboronic acid hydrochloride

A201777-1G A201777-5G
103817-328EA 8 USD
103817-328 103847-968
4-Aminophenylboronic acid hydrochloride 95%
4-Aminophenylboronic acid hydrochloride

Caution: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₆H₉BClNO₂
MW: 173.41 g/mol
Melting Pt: 195…200 °C
Flash Pt: 168.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03001333
CAS Number: 80460-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR