3-(Benzyloxy)phenylboronic acid 98%

Supplier: Ambeed

Warning

Synonyms: (3-(Benzyloxy)phenyl)boronic acid3-Benzyloxybenzeneboronic acid

A154175-5G
103842-792EA 15 USD
103842-792
3-(Benzyloxy)phenylboronic acid 98%
3-(Benzyloxy)phenylboronic acid

Caution: Keep in a dark place, sealed in dry, room temperature

Formula: C₁₃H₁₃BO₃
MW: 228.06 g/mol
Boiling Pt: 428.1 °C (760 mm Hg)
Melting Pt: 125…130 °C
Storage Temperature: Ambient
MDL Number: MFCD02093063
CAS Number: 156682-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR