2-Fluoro-3-methylphenylboronic acid 96%

Supplier: Ambeed

Warning

Synonyms: (2-Fluoro-3-methylphenyl)boronic acid

A875291-5G
103841-298EA 58 USD
103841-298
2-Fluoro-3-methylphenylboronic acid 96%
2-Fluoro-3-methylphenylboronic acid

Caution: Inert atmosphere, 2…8 °C.

Formula: C₇H₈BFO₂
MW: 153.95 g/mol
Boiling Pt: 290.5 °C (760 mm Hg)
Flash Pt: 129.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06409950
CAS Number: 762287-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR