2-Hydroxydimetridazole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1-Methyl-5-nitro-1H-imidazol-2-yl)methanol
A100321-1G
A100321-5G
A100321-25G
A100321-5G
A100321-1G
A100321-100G
A100321-250MG
77135-424EA
309
USD
77135-424
77135-420
77135-422
103821-068
103795-366
2-Hydroxydimetridazole 98%
2-Hydroxydimetridazole
Caution: for Research Use Only
Formula:
C₅H₇N₃O₃ MW: 157.1274 g/mol Boiling Pt: 422.2 °C (760 mm Hg) Melting Pt: 115…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00159675 CAS Number: 936-05-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...