Masterflex® Union Fittings, Hose Barb, Y-Connector, Polyproylene, Cole-Parmer
Supplier: Cole Parmer
06295-40 06295-50 06295-30 40726-47
MSPP-EW4072647PK 53 USD
MSPP-EW4072647 MSPP-EW0629540 MSPP-0629550 MSPP-WW0629530
Masterflex® Union Fittings, Hose Barb, Y-Connector, Polyproylene, Cole-Parmer
Fittings and Connectors
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match unique application
  • Designed for quick and easy assembly with flexible tubing
Combine these simple, economical fittings with flexible tubing for use in low pressure applications. Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, or food processing.