Lab Companion Reciprocal Shaking Water Baths, Cole Parmer
Supplier: Cole Parmer
12054-20
MSPP-AAH44211KEA 5905.38 USD
MSPP-AAH44211K
Lab Companion Reciprocal Shaking Water Baths, Cole Parmer
Baths Shaking Water Baths
PID temperature and speed control at economical prices.
  • Gentle start to the shaking action minimzes shock to samples
  • Dual LED displays show actual or set temperature and shaking speed
  • Included gable cover minimizes fluid loss from evaporation
These shaking baths feature microprocessor temperature control with built-in calibration. Store up to three temperature settings for fast recall. Shaker mechanism has its own control key enabling you to use the unit as a constant temperature bath. Digital timer can be set from 1 to 99 h 59 min and includes a delayed on/off capability. Shaking carriage can be removed allowing you to attach racks without contacting the fluid. Safety features include independent overtemperature and low-level alarms.