BioVision’s Lentivirus qPCR Quantification Kit is a one-step assay by qPCR which does not require additional lysis or reverse transcription (RT) steps, and can be completed under one hour. Our 2X qPCR MasterMix has intrinsic reverse transcriptase activity, thus eliminating the need for an extra reverse transcription incubation step.
- Reliable and ready to use
- Minimal non-specific background
- High specificity and sensitivity
- Results ready in less than 1 h
The kit is designed to deliver high sensitivity and ensures minimal non-specific background. ROX reference dye is provided separately from the MasterMix, making this kit universally compatible with most qPCR instruments.
Packaging: Ships in Dry Ice.