Entrance Sign, 'STOP DO NOT VISIT IF YOU HAVE THE FOLLOWING SYMPTOMS…' Accuform®
Supplier: Accuform
MGNG560VA MGNG560VS MGNG560VP MGNG561VA MGNG561VS MGNG561VP
76442-102EA 19.34 USD
76442-102 76442-104 76442-106 76442-096 76442-098 76442-100
Entrance Sign, 'STOP DO NOT VISIT IF YOU HAVE THE FOLLOWING SYMPTOMS…' Accuform®
Signs
Signs inform those to keep healthy, safe, and how to help prevent the spread of COVID-19, the Coronavirus.
  • Multiple material and size options
Choose from three material options. Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. Moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. Plastic signs are durable with slight flex. Good material choice for indoor and sheltered outdoor environments. The printed surface has a clear UV protective overcoat. Aluminum signs provide the strength of metal without rusting. Printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance.