Nitrile Examination Gloves, 4.8 g, Sri Trang
Supplier: SRI TRANG GLOVES (THAILAND)
14009648 14009649 14009650 7025056 14009651
76459-454CS 277.8 USD
76459-454 76459-456 76459-458 76459-452 76459-460
Nitrile Examination Gloves, 4.8 g, Sri Trang
Gloves
Used in laboratories worldwide.
  • Non sterile
  • Latex-free
  • Minimum thickness of 2.0 mil
  • Powder-free
  • 9" length
  • Medical grade
  • Finger textured with beaded cuff
While it can be used as a medical examination and category III protective glove, this latex-free protection comes in handy while working on glass equipment, in laboratories, or in kitchens. The finger texturing allows optimal performance through a high sensitivity feel throughout the glove film. The finished surface allows grabbing of any smooth and wet surface without losing grip, while protecting from any surroundings that should not come in contact with bare skin.
Certifications: ASTM D6124, ASTM D412/ ASTM D573, ASTM D3767, ASTM D6319. The gloves are manufactured in compliance with ISO 9001, ISO 13485.
Packaging: Packed 10 boxes per case.
Caution: Keep storage area cool, dry and dust free, avoid ventilation and storage close to photocopy equipment. Copper ions discolour the glove. Protect gloves against ultraviolet light sources, as sunlight and oxidizing agents. Storage above 38 °C (100 °F) will lead to accelerated aging and should be avoided under any circumstances. Long term storage in bulk can lead to pleats, stickiness and early aging of the glove and should be avoided.