Abotech N95 Mask, V-Fold, NIOSH Approved, Hardy Diagnostics
Supplier: Hardy Diagnostics
20180016
76458-110PK 120.63 USD
76458-110
Abotech N95 Mask, V-Fold, NIOSH Approved, Hardy Diagnostics
Respirators Disposable Respirators Particulate Respirators
N95 Mask with a V-Fold beginning from the nose, running to the chin to create a breathable space.
  • Fabric is durable and NIOSH approved
  • Soft silicone band on the perimeter for a comfortable seal
  • Two adjustable straps along the ears and neck
Masks feature a soft silicone band on the perimeter to create a comfortable facial seal, along with two adjustable straps along ears and neck for a secure fit.
Packaging: Masks are packaged ten per box.