Mark3 Moisture Analyzers, Sartorius
Supplier: Sartorius
LMA110-SET-P LMA110-SET LMA110-SET LMA100-SET-P LMA110-SET-P LMA100-SET-P
76412-324EA 8545.82 USD
76412-324 CA76412-322 76412-322 76412-318 CA76412-324 CA76412-318
Mark3 Moisture Analyzers, Sartorius
Analyzers Moisture Analyzers
Mark3 Moisture Analyzers for every application. The principle of operation of the Mark3 Moisture Analyzers is loss on drying. The instrument finalizes on a programmed rate of weight change compared to the actual initial weight of the sample over a programmed window of time. They feature a built-in temperature and weight calibration function to ensure advanced moisture analysis. The modular Mark3 with its add-ons of up to four heating units provides an unequaled flexibility and enables even parallel measurements of different samples.
  • Economical and reliable alternative to the Karl-Fischer titration and drying oven methods
  • Intuitive operation – With a learning curve of a few minutes
  • The user starts the measurement by pressing just one button
  • Robust and fast, optimal for routine operation
  • Multi-module operation enables parallel measurements of up to 4 different samples on one single device
  • An extensive range of security settings guarantees reliable results and 100% traceability
Mark3 HP Moisture Analyzer
A flexible moisture analyzer system for labs and production facilities. Its optional modular add-ons provide the best solution for every measuring task. Its high-resolution weighing system also makes the Mark3 HP suitable for precisely analyzing the moisture and water content of very dry samples. This feature makes it a genuine alternative to Karl-Fisher titration.
Mark3 LTE Moisture Analyzer
Fast and accurate moisture or solids measurement with the Mark3 LTE allows adjustments to be made in the processing cycle in minutes rather than hours. With its resolution of 0.01% the Mark3 LTE is an economical and reliable alternative to the drying oven and achieves excellent reproducibility in routine measurements because of its built-in Sartorius weighing technology.