Potassium iodide ≥99.0%, crystalline powder, BAKER ANALYZED® ACS, J.T.Baker®
|
Danger
|
Synonyms:
Dipotassium diiodide
, Kali iodide
Management of Change (MOC) category = N
BAKR3164-69
3164-69
JT3164-69CS
0.21
USD
CAJT3164-69
JT3164-69
Potassium iodide ≥99.0%, crystalline powder, BAKER ANALYZED® ACS, J.T.Baker®
Potassium iodide
Formula:
Kl MW: 166 g/mol Melting Pt: 681 °C Density: 3.10 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011405 CAS Number: 7681-11-0 EINECS: 231-659-4 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (KI) | ≥99.0 % |
ACS - Appearance (white crystalline powder) | Passes Test |
Identification (Potassium) | Passes Test |
Identification (Iodide) | Passes Test |
ACS - Insoluble Matter | ≤0.005 % |
ACS - Loss on Drying at 150°C | ≤0.2 % |
ACS - pH of 5% Solution at 25°C | 6.0 - 9.2 |
ACS - Chloride and Bromide (as Cl) | ≤0.01 % |
ACS - Phosphate (PO₄) | ≤0.001 % |
ACS - Sulfate (SO₄) | ≤0.005 % |
ACS - Barium (Ba) | ≤0.002 % |
ACS - Calcium (Ca) | ≤0.002 % |
ACS - Magnesium (Mg) | ≤0.001 % |
ACS - Sodium (Na) | ≤0.005 % |
Iodate (IO₃) | ≤3 ppm |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤3.000 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...