Universal Transport Medium, Azer Scientific
Supplier: Azer Scientific
PFUTM-10
76441-350CS 2714.65 USD
76441-350
Universal Transport Medium, Azer Scientific
Media for Microbiology Prepared Media for Microbiology
The universal transport medium consists of Hank's balanced salt solution modified and enriched with bovine serum albumin, amino acid, gelatin peptone and carbohydrate. The pH is adjusted with buffer.
  • For the collection and preservation of virus, chlamydia, mycoplasma and ureaplasma
  • Tested and validated for excellent recovery and viability of a variety of viruses, including SARS-CoV-2
  • Room temperature stable