NativeFolder Bacterial Culture Medium 1L 2
Supplier: MOLECULAR DEPOT LLC MS
K1010004
MSPP-K1010004EA 403.85 USD
MSPP-K1010004
NativeFolder Bacterial Culture Medium 1L 2
Cell Culture Media
NativeFolder is a bacterial culture medium specially formulated to promote the folding and the expression of soluble recombinant proteins in E. coli.
NativeFolder contains proprietary compounds that prevent protein aggregation and misfolding during protein expression. Don’t waste your time experimenting with the renaturation of inclusion bodies. Use NativeFolder and express your correctly folded target protein in the soluble fraction. Simply add water to the provided mix, autoclave and filter sterilize. Recombinant proteins of up to 110 kDa were successfully expressed as soluble fractions using NativeFolder.