Magid® Reusable Cloth Face Masks, Magid Glove
BARRIERMASK
76417-628BX 408.33 USD
76417-628
Magid® Reusable Cloth Face Masks, Magid Glove
Masks are constructed from a double-layer material, with 5.5" interior pocket for filter media and two ear straps.
  • Reusable non-medical face mask
  • Designed according to the CDC’s recommended cloth face mask guidelines
  • Allows for breathing without restriction
  • Fits snugly but comfortably against the side of the face with two braided elastic ear loops
  • Double-layer woven polyester material with an interior pocket for filter media
  • Machine washable and reusable
These masks are not medical grade. Face masks do not supply oxygen.
Ordering information: Orders cannot be returned.