Trihalomethanes Volatiles Mix (High Level) for US EPA Method 502.2
Supplier: Avantor
VWR Chemicals BDH®
BDH0805-1ML
BDH0805-1MLEA 62.2 USD
BDH0805-1ML
Trihalomethanes Volatiles Mix (High Level) for US EPA Method 502.2
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 502.2 is a general analytical method for the monitoring of halogenated, purgeable VOCs in drinking water and raw water sources by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).