BIOne 1250 Bioprocess Control Station with Autoclavable Bioreactor for Microbial Models, Distek
2022-8100-R5AM 2022-8100-M5AM 2022-8100-M2AM 2022-8100-R2AM
76419-888EA 68632.61 USD
76419-888 76419-902 76419-896 76419-882
BIOne 1250 Bioprocess Control Station with Autoclavable Bioreactor for Microbial Models, Distek
Bioreactors
Distek, Inc. has revolutionized benchtop scale bioreactors with the release of our BIOne 1250 bioprocess control station with autoclavable bioreactor for microbial models. The sleek appearance and intuitive touchscreen interface are only the beginning. The BIOne 1250 is designed to be the easiest to use and most capable stand-alone controller on the market.
  • Touchscreen interface
  • Smart media filling
  • Email and text alerts
  • Trend up to eight parameters
  • Variable speed pumps
  • 2, 5 and 10 L working volumes
  • Configurable for your lab
Control all bioreactor parameters directly from the BIOne 1250 interface, or, for remote control, use our optional OPC server and communicate with your choice of SCADA system.
From the pop-up tab of your selected parameter, control setpoints, calibrations, alarms, and more. Reduce development timelines by utilizing the live, dual graph trend screen. Analyze and compare current and previous runs. Track all optimized changes and export data to further analyze.
Certifications: ETL, CE
Ordering information: Includes touchscreen display, vessel, headplate, stand, heater jacket, 4 pumps, pH & DO sensors and accessory kit. Includes a 1 year comprehensive warranty. Does not include; installation or qualification services.