KN95 Respirator Masks, Wells Lamont
Supplier: WELLS LAMONT INDUSTRY GROUP
WLIKN95CB
76414-934BX 267.8 USD
76414-934
KN95 Respirator Masks, Wells Lamont
Respirators Disposable Respirators Particulate Respirators
Protect employees and prevent contamination with this disposable KN95 respirator masks.
  • KN95 mask filters out 95% of 0.3 µm particles
  • Affixes securely and comfortably with ear loops
Creates an airtight seal around the face. The ear loops make this KN95 mask easier to apply and remove, and ensure a more comfortable, long-term wear than traditional N95 masks.
Certifications: FFP2 Standard- EN 149:2001 + A1:2009
Packaging: Masks are packaged 50 per box.