Pistons and Seals, Agilent Technologies
Supplier: Agilent Technologies
G5611-60016 G4220-60016 5063-6589 G4204-40005 G1376-60015 5063-6586 5067-4695 G1361-22402 5067-5678 0905-1420 G4220-60015 5022-2188 5067-5975 0905-1719
101389-492EA 321.32 USD
101389-492 103582-704 101389-556 103577-236 101389-554 101471-900 101479-952 101401-456 101401-468 101481-854 101484-380 101481-856 101449-220 101479-488
Pistons and Seals, Agilent Technologies
These pistons are made from a high purity, mono-crystalline sapphire for maximum durability.
  • Consistent, reproducible results
Although ceramic pistons can be manufactured at a lower cost, ceramic is a sintered, polycrystalline material, which can cause undesirable variations during the production process. Agilent seals are designed to fit snugly around our pistons, and are capable of adapting to a wide range of flow rates and pressures. The combination of our piston and seal has undergone extensive testing under temperature stress, with all common HPLC solvents, and in many instruments.
Agilent sapphire pistons are: Meticulously cut at just the right angle, making them durable – and long lasting. Precisely aligned in their stainless steel holder to minimize wear on the piston and seal.
Agilent piston seals are: Spring-loaded and engineered to deliver optimal performance over highly dynamic flow and pressure ranges. Manufactured from a proprietary polymer blend, and feature a spring made from the same high-quality stainless steel that is used in our pump's flow path.