Brady® COVID-19 Signs; Please Sanitize After Use, Brady Worldwide
Supplier: Brady
170555 170552 170554 170553 170551 170556
76420-508EA 21.65 USD
76420-508 76420-506 76420-500 76420-510 76420-504 76420-502
Brady® COVID-19 Signs; Please Sanitize After Use, Brady Worldwide
Signs
Having the right information helps keep everyone safe. Post this sign in the facility to keep employees and visitors informed on the best ways to prevent the spread of COVID-19.
  • Signs are available in B-302 polyester, B-555 aluminum, or B-401 polystyrene
  • B-302 polyester signs are self adhesive
  • B-555 aluminum and B-401 polystyrene signs feature corner holes for mounting and placement
B-302 polyester is an indoor/outdoor high-performance material with a permanent pressure-sensitive adhesive. This flexible material conforms to most surfaces and provides clear identification in even the harshest environments. It survives drastic temperature variations up to 230° F (110° C), and can be applied at 0° F (-18° C).
Durable non-reflective B-555 aluminum material provides lasting identification with durable inks and clear-coat protection for added fade resistance. It is designed for both indoor and outdoor use.
B-401 polystyrene material is ideal for use in light-duty areas and is perfect for temporary identification. This temporary sign provides excellent abrasion and chemical resistance, with a service temperature of -40 to 175° F (-40 to 80°C).