Brady® COVID-19 Signs; Temperature Screening, Bi-lingual, Brady Worldwide
Supplier: Brady
170537 170536 170538 170534 170535 170539
76420-470EA 14.43 USD
76420-470 76420-480 76420-474 76420-472 76420-478 76420-476
Brady® COVID-19 Signs; Temperature Screening, Bi-lingual, Brady Worldwide
Signs
Having the right information helps keep everyone safe. Post this sign in the facility to keep employees and visitors informed on the best ways to prevent the spread of COVID-19.
  • Signs are available in B-302 polyester, B-555 aluminum, or B-401 polystyrene
  • B-302 polyester signs are self adhesive
  • B-555 aluminum and B-401 polystyrene signs feature corner holes for mounting and placement
B-302 polyester is an indoor/outdoor high-performance material with a permanent pressure-sensitive adhesive. This flexible material conforms to most surfaces and provides clear identification in even the harshest environments. It survives drastic temperature variations up to 230° F (110° C), and can be applied at 0° F (-18° C).
Durable non-reflective B-555 aluminum material provides lasting identification with durable inks and clear-coat protection for added fade resistance. It is designed for both indoor and outdoor use.
B-401 polystyrene material is ideal for use in light-duty areas and is perfect for temporary identification. This temporary sign provides excellent abrasion and chemical resistance, with a service temperature of -40 to 175° F (-40 to 80°C).