Defender Face Shield, Troemner
Supplier: Troemner
30596444
76415-652CS 157.99 USD
76415-652
Defender Face Shield, Troemner
Face Shields
The Troemner Defender face shield combines exceptional comfort (lightweight, impregnated anti-fog design, and foam pad for comfort and airflow) and safety (latex-free, protects face and neck).
  • Ergonomic design: lightweight, comfortable forehead pad
  • Protects against airborne pathogens, liquid/solid samples
  • Offers distortion-free visibility (central and peripheral vision)
  • Compatible with other PPE (goggles, safety glasses, mask)
  • Latex-free
  • Disposable, easy to clean
  • Anti-fog design: open sides and anti-fog impregnated, not coated
  • Anti-static on both side
The disposable face shield is intended to protect healthcare providers who are in close contact with humans, samples (aerosols, biofluids, biosolids), and other biohazardous materials or dangerous chemicals.