Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • Environmentally Preferable (22) GSA Contract (1) MWBE (32) SBE (31)
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

Cloths for General Applications

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"77"
Sort by:
 
 
 
 

Catalog Number: (21902-985)
Supplier:  VWR International
Description:   Soft, pure, highly absorbent pads will not scratch the most delicate coatings or surfaces.
Product available on GSA Advantage®
Catalog Number: (21902-990)
Supplier:  VWR International
Description:   This non-woven material is a unique, high-quality fabric made specifically for the Graphic Arts Industry.
Catalog Number: (470152-954)
Supplier:  VWR
Description:   Cloth, Cotton & Terry, 6 x 12" 1/Pkg
Supplier:  Kimberly-Clark
Description:   Durable, uniformly-reinforced towel. Embossed surface traps and holds dirt and liquids. Can be used in a full range of medium- to heavy-duty applications. No adhesives or binders. Color: white. Size: 23.1x42.7cm. POP-UP* dispenser box of 126 wipers.
Supplier:  Kimberly-Clark
Description:   For heavy manufacturing, refining, rubber/plastics, metal fabrication, machine, tool/die shops. Made of fast-absorbing HYDROKNIT* hydroentangled cellulose/polypropylene blend. Durable. Reusable. Color: White. Size: 31.8x36.6cm. Packaged 76 quarter-fold rags per box.
Supplier:  Vileda Professional - FHP
Description:   Microfiber cloth with very finely split microfibers for superior cleaning performance
Supplier:  Kimberly-Clark
Description:   Pound for pound, absorb more water and oil than rental shop towels. SuperCharged technology adds softness plus absorbency. 12.5" x 14.4", 50 towels per pack, 4 packs per case. White. 1/4 fold.
Supplier:  Kimberly-Clark
Description:   KIMTECH PREP® KIMTEX™ Wipers feature 100% polypropylene construction for cloth-like feel and durability
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 77
  1  2  3  4  5  6  7  8  9  10  Next