Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • Environmentally Preferable (14) GSA Contract (70) SBE (55) SBE (4)
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "laboratory-coats"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"125"
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   These lab coats are the cleanest and most durable available within the VWR® Protection line.
Supplier:  VWR International
Description:   Fashionable lab coats for women feature a tapered back panel with a half belt. Lab coats are manufactured from durable and comfortable poly/cotton (80...
Supplier:  VWR International
Description:   VWR® Protection ESD Lab Coats are manufactured from breathable and comfortable 98% cotton fabric and 2% carbon fabric. These ESD Lab Coats provide exc...
Supplier:  VWR International
Description:   Frocks are manufactured from a multilayered spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength while remaining soft, comfo...
Supplier:  VWR International
Description:   Unisex Technology lab coats are designed for use in research environments, quality-control laboratories, and industrial areas.
Supplier:  VWR International
Description:   Frocks provide a high level of protection in cleanroom environments. Garments are chemically and biologically inert, low-linting, and static-free unti...
Supplier:  VWR International
Description:   Irradiated frocks are manufactured from a specially formulated breathable microporous fabric that is designed to provide significant fluid and barrier...
Supplier:  VWR International
Description:   These are the cleanest and most durable frocks available within the VWR® Protection line.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 125
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next