Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

Safety Boots

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"28"
Sort by:
 
 
 
 

Supplier:  Onguard Industries
Description:   100% waterproof, high molecular weight
Supplier:  Superior Uniform
Description:   Reusable, high-top cleanroom boots have a flexible Hypalon® sole with upper of static dissipative Burlington C3™ material
Supplier:  Honeywell Safety Products
Description:   Seamless, waterproof boots feature Triple Density Technology® (TDT®), a three-stage molded design that can withstand harsh environments with slick or ...
Supplier:  PORTWEST LLC.
Description:   The Total Safety Wellington offers ultimate S5 protection to the wearer
Supplier:  Honeywell Safety Products
Description:   16 inch knee boot. SAFE-TOE steel toe construction. Anti-Skid sole and heel. FOOT FORM contour insole. Made in U.S.A. Size: 11
Supplier:  Onguard Industries
Description:   Size 10. Two-stage, injection-molded, one-piece construction. Waterproof. Polyurethane/PVC. Non-slip soles. Pull-up/kick-off lugs. Steel shank. Removable cushion insole. Meets or exceeds ANSI Z41 PT91 I/75 C/75 for impact and compression. CSA Z195 M92 Grade 1 certified.
Catalog Number: (470004-694)
Supplier:  TRIPLE S SPORTING SUPPLIES INC
Description:   The wade is over - the way to look Hip, even in water up to your chest!
Limited Availability
Supplier:  PORTWEST LLC.
Description:   Welted Safety Boots with steel toecap and steel midsole
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 28
  1  2  3  4  Next