Freestanding and Bolt-on Breath Shields, Eagle MHC
Supplier: Eagle MHC
YJ-4286-00 YJ-4278-00 YJ-4285-00 YJ-4256-00 YJ-4260-00
76416-062EA 262.5 USD
76416-062 76416-058 76416-060 76416-064 76416-056
Freestanding and Bolt-on Breath Shields, Eagle MHC
Shields Safety Shields
Add a layer of protection to help maintain social distancing and provide a physical barrier to protect staff and customers.
  • Durable clear plexiglass protection
  • Uniform 0.218" thickness
  • Multiple sizes and styles are available
Eagle Breath Shields are available in both freestanding and bolt-on formats to provide a physical barrier to protect both workers and customers. Freestanding breath shields allow for portable, flexible placement that can be adjusted based on specific needs.
These shields feature a pass-through cutout and require no tools for assembly. Bolt-on shields allow users to retrofit counters and workstations to conform to new protective requirements. These shields can be easily mounted to the front of a counter, and require only three bolts or screws for secure application.