Quartz Baffled Cyclonic Spray Chamber without Gas Port for ELAN® 5000/6x00/9000/DRCs PC3 Systems, PerkinElmer
Supplier: PerkinElmer
N0777035
10138-200EA 2649.68 USD
10138-200
Quartz Baffled Cyclonic Spray Chamber without Gas Port for ELAN® 5000/6x00/9000/DRCs PC3 Systems, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
This quartz baffled cyclonic spray chamber, with drain line, auxiliary gas port, and injector connection, is used in conjunction with PC3 Peltier cooler inlet systems with ELAN® 5000, 6x00, 9000, and DRC variations ICP-MS series instruments.
The quartz baffled cyclonic spray chamber features a 7 mm baffle and auxiliary gas port. It's designed for all-purpose, non-hydrofluoric acid-resistant applications, and provides excellent sensitivity and stability with low background.