Reducer Fittings, PP, Cole-Parmer®
Supplier: Cole Parmer
06365-40
76377-402PK 129.27 USD
76377-402
Reducer Fittings, PP, Cole-Parmer®
Connectors Connectors, Tubing
These polypropylene straight barbed reducing fittings easily connect fluid paths without the use of tools. Simply slip your flexible tubing or hose onto the barb to create a secure seal.
  • Best for low-pressure applications
Ideal for general-purpose and single-use applications, to connect tubing with rest of system.