The G450 gas detector offers exceptional performance for an extremely affordable price. The four sensor multi-gas detector is compact, durable, and very easy to use and are warranted for three full years, ensuring that it has the lowest cost-of-ownership of any confined space gas detector on the market today.
- Interchangable battery packs for up to 24 hours of continuous operation
- Standard event and datalogging
- Highly water resistant (IP 67)
- Optional motorized smart pump operates in diffusion or sample-draw mode
The H₂S sensor offers 0.1 ppm resolution to comply with the latest TLV requirements.
It offers completely automatic calibration, one-button operation, top mounted display and interchangeable battery packs for up to 24 hours of continuous operation.
The optional motorized pump operates in diffusion and sample-draw modes, and can be easily attached or removed from the instrument when sampling needs change.
Certifications: CSA Certified, MSHA certified instruments available.
Accessories information: Optional items – pump, 10' tubing, probe, datalogging kit, docking station, test station, Cal gas, kits, regulators.
Ordering information: Replacement sensors carry the same three year warranty as the original sensors. For available accessories or additional information, contact your VWR representative and see the attached datasheet and manual.
Delivery information: Standard items included are G450 detector, specified sensors, charger (if specified), calibration adapter, manual, CD.
Value kit: (if specified), all standard items, Cal gas, fixed flow regulator, ABS case, hand aspirated or motorized pump.
Confined space kit: (if specified), all value kit items, aluminum case, spare alkaline battery, hand aspirated or motorized pump, data downloading software, cable pump filters.