Avantor® ACE® SuperC18, HPLC Columns, 3 µm
Supplier: VWR
Avantor® ACE® SuperC18
ACE111115003S
76384-292EA 26434.39 USD
76384-292
Avantor® ACE® SuperC18, HPLC Columns, 3 µm
Chromatography Columns
Avantor® ACE® SuperC18™ columns are specially designed for high and low pH mobile phases, for use with LC/MS compatible buffers. These columns are highly resistant to ligand cleavagee, due to a combination of the Encapsulated Bonding Technology (EBT™).
  • Recomended pH range: 1.5 to 11.5
  • Compatible with both MeOH and MeCN
  • Rapid column equilibration without memory effects
  • Offers ultra-low bleed for improved LC-MS compatibility
  • Stainless steel column hardware
Avantor® ACE® SuperC18™ column is ideal choice for method development includes exploit selectivity changes at low, intermediate and high pH, excellent column lifetime for reduced analysis costs.