Avantor® Hichrom, Titanium Frit
Supplier: VWR
Avantor® Hichrom
HI-112
76399-128EA 23.78 USD
76399-128
Avantor® Hichrom, Titanium Frit
Filters Precolumn Filters and Frits
Titanium frits are resistant to corrosion, biocompatible and recommended for protein analysis.