Autosampler Sample Pans, 50 µl, PerkinElmer
Supplier: PerkinElmer
B0169321
10136-336EA 2731.65 USD
10136-336
Autosampler Sample Pans, 50 µl, PerkinElmer
Trays/Dishes/Pots Laboratory Trays/Dishes
These sample pans have a volume of 50 µL. The Autosampler sample pans are used to prepare sample materials for DSC Autosampler.
  • Pans are available in sealed or vented versions
  • Used for non-robotic systems
  • Can be used with all autosamplers
These sample pans are for use with Diamond DSC, Pyris 1 DSC, DSC 7, DSC 6, Pyris 6 DSC, and Jade DSC.
Delivery information: Includes 400 sample pans.