INTERCEPT™ technology ANSI cut level 4 performance for the ultimate in cut resistance.
- Glass-free INTERCEPT™ liner provides increased cut protection
- Silicone and DMF-free
- Grip performance for safe handling in dry to light oily applications
- Double dipped nitrile coating protects against oil and industrial fluid exposure
- INTERCEPT™ Aramid Liner for increased cut performance
- Contact Heat Protection level 1
The glove is composed of double layer RIPEL™ nitrile coating, which keeps hands dry and comfortable when handling oily parts and tools. The HyFlex® 11-928 also offers intermittent contact hear protection when handling hot parts up to 212 °F.
Certifications: ANSI 3 ABR; ANSI A4 CUT; EN 420:2003 + A1:2009; Category lll; REACH compliant.
Packaging: Gloves are packaged 12 pairs per polybag, 12 polybags per carton.