WHEATON® Dropper Bottles, DWK Life Sciences
Supplier: DWK Life Sciences
W009912
10027-478CS 2418.22 USD
10027-478
WHEATON® Dropper Bottles, DWK Life Sciences
Bottles Dropping and Dispensing Bottles
White finish for UV light-sensitive products.
  • Dispenses reliable, repeatable uniform drops
  • Excellent alternative to pipetting
  • Manufactured to rigid ISO and quality standards
  • Excellent chemical resistance
  • Ideal for most biotech, diagnostic, and pharmaceutical applications
Friction fit tips are available in a wide variety of colors. Products are double bagged and twist-tied to reduce particulates. Methods of sterilization are gas and gamma.
Ordering information: Custom configurations are available upon request.