KIMBLE® Freeze Drying Hose Adapters, DWK Life Sciences
Supplier: DWK Life Sciences
562850-0012 562850-0516
KT562850-0012EA 67.22 USD
KT562850-0012 KT562850-0516
KIMBLE® Freeze Drying Hose Adapters, DWK Life Sciences
These adapters are intended for connection to vacuum hose and a KT562830-0000 vacuum adapter.
  • ¹/₂" (12.7 mm) ID size will fit ³/₄" (19.1 mm) ID hose
  • ⁵/₁₆" (7.9 mm) ID will accept ¹/₂" (12.7 mm) ID hose
Delivery information: Supplied with a size 018 FKM O-ring.