BD BBL™ Inhibitory Mold Agar with Chloramphenicol and Gentamicin, Prepared Media Stacker™ Plates, BD Diagnostics
Supplier: BD Diagnostics
BD BBL™
299715
90005-258EA 49.82 USD
90005-258
BD BBL™ Inhibitory Mold Agar with Chloramphenicol and Gentamicin, Prepared Media Stacker™ Plates, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Inhibitory mold agars provide enhanced selectivity for the isolation of pathogenic and nonpathogenic fungi from a variety of clinical and nonclinical specimens and come in 100×15 mm petri-style configurations.
  • Ready for immediate use
Selectivity is further enhanced by providing the medium, inhibitory mold agar with chloramphenicol and gentamicin, with double the concentration of chloramphenicol contained in inhibitory mold agar base. The plates are deep-filled to reduce the effects
of drying during prolonged incubation.